missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
![](http://beta-static.fishersci.ca/images/Gray.gif)
Sodium Acetate Trihydrate (Crystalline/Certified ACS), Fisher Chemical™
$272.25 - $9802.17
Chemical Identifiers
CAS | 6131-90-4 |
---|---|
Molecular Formula | C2H9NaO5 |
Molecular Weight (g/mol) | 136.079 |
MDL Number | MFCD00071557 |
InChI Key | AYRVGWHSXIMRAB-UHFFFAOYSA-M |
Synonym | sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn |
PubChem CID | 23665404 |
ChEBI | CHEBI:32138 |
IUPAC Name | sodium;acetate;trihydrate |
SMILES | CC(=O)[O-].O.O.O.[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
S209-500
![]() |
Thermo Fisher Scientific
S209500 |
500 g | Poly Bottle |
Each for $272.25
Case of 6 Each for $1,633.50
|
![]() |
||||
S209-3
|
Thermo Fisher Scientific
S2093 |
3 kg | Poly Bottle |
Each for $772.92
Case of 4 Each for $3,091.68
|
|
||||
S209-10
|
Thermo Fisher Scientific
S20910 |
10 kg | Poly Pail |
Each for $2,156.92
|
|
||||
S20950
|
Fisher Chemical
FLS20950 |
50 kg | Fiber Drum |
Each for $9,802.17
|
|
||||
Chemical Identifiers
6131-90-4 | |
136.079 | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
23665404 | |
sodium;acetate;trihydrate |
C2H9NaO5 | |
MFCD00071557 | |
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
CHEBI:32138 | |
CC(=O)[O-].O.O.O.[Na+] |
Specifications
58°C | |
White | |
Solid | |
99.0 to 101.0 % | |
CH3COONa·3H2O | |
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
CC(=O)[O-].O.O.O.[Na+] | |
136.079 | |
CHEBI:32138 | |
99.0 to 101.0% | |
Pass Test | |
1.45g/cm³ |
6131-90-4 | |
8 to 9.5 | |
500 g | |
C2H9NaO5 | |
MFCD00071557 | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
sodium;acetate;trihydrate | |
23665404 | |
136.08 | |
Certified ACS | |
Poly Bottle | |
Sodium Acetate Trihydrate |
Safety and Handling
![](http://beta-static.fishersci.ca/images/Gray.gif)
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:1
Instability:1