missing translation for 'onlineSavingsMsg'
Learn More

Sodium amide, 99%, extra pure

CAS: 7782-92-5 | H2NNa | 39 g/mol

$102.81 - $102.81

Chemical Identifiers

CAS 7782-92-5
Molecular Formula H2NNa
Molecular Weight (g/mol) 39
MDL Number MFCD00011117
InChI Key ODZPKZBBUMBTMG-UHFFFAOYSA-N
Synonym sodium amide, sodamide, sodium amide na nh2, unii-5db3g6px9d, 5db3g6px9d, sodium azanide, sodium amide, 50 wt. % suspension in toluene, sodiumamide, natriumamid, sodioamine
PubChem CID 24533
IUPAC Name sodium;azanide
SMILES [NH2-].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC339240250
SDS View Documents
Thermo Scientific Chemicals
339240250
25 g Glass bottle
Each for $102.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7782-92-5
39
ODZPKZBBUMBTMG-UHFFFAOYSA-N
24533
[NH2-].[Na+]
H2NNa
MFCD00011117
sodium amide, sodamide, sodium amide na nh2, unii-5db3g6px9d, 5db3g6px9d, sodium azanide, sodium amide, 50 wt. % suspension in toluene, sodiumamide, natriumamid, sodioamine
sodium;azanide

Specifications

400.0°C
7782-92-5
Powder, Irregular Flakes or Granules
99%
NaNH2
01,1034; 02,373; 04,439; 05,591; 06,525; 08,446; 15,288
sodium amide, sodamide, sodium amide na nh2, unii-5db3g6px9d, 5db3g6px9d, sodium azanide, sodium amide, 50 wt. % suspension in toluene, sodiumamide, natriumamid, sodioamine
ODZPKZBBUMBTMG-UHFFFAOYSA-N
sodium;azanide
24533
99%
Glass bottle
210.0°C
Gray-White to White
25 g
H2NNa
MFCD00011117
15, 8714
Solubility in water: reacts violently with water. Other solubilities: decomposes in hot alcohol
[NH2-].[Na+]
39
39
Extra Pure
Sodium amide, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Very toxic to aquatic life.
Reacts violently with water.
May form explosive peroxides.<

GHS P Statement
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Danger

EINECSNumber : 231-971-0

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only