Learn More
Sodium azide, 99%, extra pure
CAS: 26628-22-8 | N3Na | 65.01 g/mol
$38.70 - $384.64
Chemical Identifiers
| CAS | 26628-22-8 |
|---|---|
| MDL Number | MFCD00003536 |
| InChI Key | PXIPVTKHYLBLMZ-UHFFFAOYSA-N |
| Synonym | sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german |
| PubChem CID | 33557 |
| ChEBI | CHEBI:278547 |
| IUPAC Name | sodium;azide |
| SMILES | [N-]=[N+]=[N-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC190380050
|
Thermo Scientific Chemicals
190380050 |
5 g |
Each for $38.70
|
|
|||||
|
AC190381000
|
Thermo Scientific Chemicals
190381000 |
100 g |
Each for $143.88
|
|
|||||
|
AC190385000
|
Thermo Scientific Chemicals
190385000 |
500 g |
Each for $384.64
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, For Azidation/Diazo transferChemical Identifiers
| 26628-22-8 | |
| PXIPVTKHYLBLMZ-UHFFFAOYSA-N | |
| 33557 | |
| sodium;azide |
| MFCD00003536 | |
| sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german | |
| CHEBI:278547 | |
| [N-]=[N+]=[N-].[Na+] |
Specifications
| 300.0°C | |
| 26628-22-8 | |
| 9 (5% min. soln.) | |
| 5 g | |
| N3Na | |
| MFCD00003536 | |
| 15, 8717 | |
| Solubility in water: 420g/L (17°C). Other solubilities: soluble in liquid ammonia, slightly soluble in alcohol, insoluble in diethyl ether | |
| [N-]=[N+]=[N-].[Na+] | |
| 65.01 | |
| CHEBI:278547 | |
| 99% | |
| 0.5% max. | |
| 1.85 | |
| 1.8500g/mL |
| 275.0°C | |
| White | |
| Crystalline Powder | |
| 98.5% min | |
| NaN3 | |
| 01,1041; 02,376; 03,259; 04,440; 05,593; 15,289; 17,68 | |
| sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german | |
| PXIPVTKHYLBLMZ-UHFFFAOYSA-N | |
| sodium;azide | |
| 33557 | |
| 65.01 | |
| Extra Pure | |
| Glass Bottle | |
| >275°C | |
| Sodium azide, 99% |
Safety and Handling
GHS H Statement:
Fatal if swallowed.
Very toxic to aquatic life with long lasting effects.
Contact with acids liberates very toxic gas.
Emergency Overview
Very toxic if swallowed. Very toxic in contact with skin. May be fatal if absorbed through skin or swallowed. Toxic by inhalation. May cause pulmonary edema. Irritating to eyes, respiratory syste
Signal Word: DANGER!
EINECSNumber : 247-852-1
RTECSNumber : VY8050000
TSCA : TSCA
RUO – Research Use Only