missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Benzoate 98.0+%, TCI America™

Supplier:  TCI America S059325G

 View more versions of this product

Catalog No. S059325G


Explore available promotions

Description

Description

Specifications

Specifications

Sodium Benzoate
532-32-1
C7H5NaO2
sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
[Na+].[O-]C(=O)C1=CC=CC=C1
144.11
144.10
25 g
450°C
White
MFCD00012463
WXMKPNITSTVMEF-UHFFFAOYSA-M
sodium benzoate
517055
≥98.0% (HPLC,T)
Crystalline Powder

Chemical Identifiers

532-32-1
144.11
WXMKPNITSTVMEF-UHFFFAOYSA-M
517055
[Na+].[O-]C(=O)C1=CC=CC=C1
C7H5NaO2
MFCD00012463
sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
sodium benzoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1272&(3)-1293

RTECSNumber : DH6650000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions