Learn More
Sodium bicarbonate, 1M buffer soln., pH 8.0, Thermo Scientific Chemicals
$154.10 - $154.10
Chemical Identifiers
CAS | 144-55-8 |
---|---|
Molecular Formula | CHNaO3 |
Molecular Weight (g/mol) | 84.01 |
MDL Number | MFCD00003528 |
InChI Key | UIIMBOGNXHQVGW-UHFFFAOYSA-M |
PubChem CID | 516892 |
ChEBI | CHEBI:32139 |
SMILES | [Na+].OC([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ62495AP
|
Thermo Scientific Chemicals
J62495AP |
500 mL |
Each for $154.10
|
|
|||||
Description
The bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents and strong acids.
Chemical Identifiers
144-55-8 | |
84.01 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
CHEBI:32139 |
CHNaO3 | |
MFCD00003528 | |
516892 | |
[Na+].OC([O-])=O |
Specifications
144-55-8 | |
8.0 | |
500 mL | |
MFCD00003528 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
sodium;hydrogen carbonate | |
516892 | |
Sodium bicarbonate buffer soln. |
1 M | |
Liquid | |
CHNaO3 | |
It is soluble in water. | |
[Na+].OC([O-])=O | |
84.01 | |
CHEBI:32139 |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only