missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bicarbonate, 99.5%, for analysis
CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
Supplier: Thermo Scientific Chemicals 217120025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferSpecifications
| Sodium bicarbonate | |
| 144-55-8 | |
| Fine Crystalline Powder | |
| 99.50% | |
| NaHCO3 | |
| 03,260; 05,595; 11,476; 17,180 | |
| sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
| UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
| sodium hydrogen carbonate | |
| 516892 | |
| 84.01 | |
| Analytical |
| 270.0°C | |
| White | |
| 2.5 kg | |
| CHNaO3 | |
| MFCD00003528 | |
| 15, 8719 | |
| Solubility in water: 9 g/100mL (20°C). Other solubilities: 16.4 g/100mL water (60°C), insoluble in alcohol | |
| [Na+].OC([O-])=O | |
| 84.01 | |
| CHEBI:32139 | |
| 99.5% | |
| Plastic bottle |
Chemical Identifiers
| 144-55-8 | |
| 84.01 | |
| UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
| 516892 | |
| [Na+].OC([O-])=O |
| CHNaO3 | |
| MFCD00003528 | |
| sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
| CHEBI:32139 |
Safety and Handling
EINECSNumber : 205-633-8
RUO – Research Use Only