missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Bicarbonate (Powder/Certified ACS), Fisher Chemical™

$223.58 - $10752.17

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
S233-500
SDS View Documents Promotion Details
Thermo Fisher Scientific
S233500
500 g Poly Bottle
Each for $223.58
Case of 6 Each for $1,341.48
 
S233-300LB
SDS Accessories View Documents
Thermo Fisher Scientific
FLS233300LB
300 lb. Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
S233-3 Edge
SDS View Documents Promotion Details
Thermo Fisher Scientific
S2333
3 kg Poly Bottle
Each for $381.25
Case of 4 Each for $1,525.00
Edge
 
S233-10
View Documents Promotion Details
Thermo Fisher Scientific
S23310
10 kg Poly Pail
Each for $1,086.08
 
S233-50
SDS Accessories View Documents Promotion Details
Thermo Fisher Scientific
S23350
50 kg Fiber Drum
Each for $4,801.92
 
Description

Description

Specifications

Chemical Identifiers

144-55-8
84.01
UIIMBOGNXHQVGW-UHFFFAOYSA-M
516892
sodium hydrogen carbonate
CHNaO3
MFCD00003528
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
CHEBI:32139
[Na+].OC([O-])=O

Specifications

270°C
White
Powder/Solid
99.7 to 100.3 %
MFCD00003528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium hydrogen carbonate
516892
84.01
Certified ACS
Poly Bottle
Sodium Bicarbonate
144-55-8
8.3
500 g
CHNaO3
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
[Na+].OC([O-])=O
84.01
CHEBI:32139
99.7 to 100.3%
Pass Test
>50°C
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Irritating to eyes and skin. May cause adverse kidney effects. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:1

SDS
missing translation for 'documents'

missing translation for 'documents'