missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate, Saturated, Ricca Chemical

$90.28 - $203.50

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
7148-32
View Documents
Ricca Chemical Company
714832
1 L Natural Poly Bottle N/A
Only null left
 
7148-1
View Documents
Ricca Chemical Company
71481
4 L Natural Poly Bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

144-55-8
84.01
UIIMBOGNXHQVGW-UHFFFAOYSA-M
516892
sodium hydrogen carbonate
CHNaO3
MFCD00003528
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
CHEBI:32139
[Na+].OC([O-])=O

Specifications

Colorless
89.0,8.91
CHNaO3
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
[Na+].OC([O-])=O
84.01
CHEBI:32139
Natural Poly Bottle
Slightly Alkaline
Liquid
144-55-8,7732-18-5
93.0,9.28
MFCD00003528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium hydrogen carbonate
516892
Laboratory
Saturated Aqueous Solution
1 L
Sodium Bicarbonate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'