Learn More
Sodium bismuth oxide, ACS, 80% min
Manganese testing in iron, steel, ores | CAS: 12232-99-4 | BiNaO3 | 279.967 g/mol
$155.18 - $460.74
Chemical Identifiers
| CAS | 12232-99-4 |
|---|---|
| Molecular Formula | BiNaO3 |
| Molecular Weight (g/mol) | 279.967 |
| MDL Number | MFCD00003474 |
| InChI Key | PNYYBUOBTVHFDN-UHFFFAOYSA-N |
| Synonym | sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium |
| PubChem CID | 4063671 |
| IUPAC Name | sodium;oxido(dioxo)bismuth |
| SMILES | [O-][Bi](=O)=O.[Na+] |
Description
Sodium bismuth oxide is used for the determination of manganese in iron, steel and ores. It is also used in the separation of plutonium and acts as a strong oxidizer. It is used in the conversion of manganese compound to permanganate. Further, it is used in the preparation of hexavalent americium (Am) during its separation from the lanthanides and in the hydrothermal synthesis of bismuth oxychloride micrometer-sized single crystals. In addition to this, it is employed as a precursor in the preparation of bismuth nanoforms.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12232-99-4 | |
| 279.967 | |
| PNYYBUOBTVHFDN-UHFFFAOYSA-N | |
| 4063671 | |
| [O-][Bi](=O)=O.[Na+] |
| BiNaO3 | |
| MFCD00003474 | |
| sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium | |
| sodium;oxido(dioxo)bismuth |
Specifications
| 12232-99-4 | |
| Powder | |
| BiNaO3 | |
| 14,8585 | |
| Insoluble in cold water; Decomposes in hot water forming Bi2O3,NaOH,and oxygenInsoluble in water. | |
| [O-][Bi](=O)=O.[Na+] | |
| 279.967 | |
| 279.97 | |
| ACS | |
| Hygroscopic |
| Yellow to Brown | |
| 25 g | |
| MFCD00003474 | |
| sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium | |
| PNYYBUOBTVHFDN-UHFFFAOYSA-N | |
| sodium;oxido(dioxo)bismuth | |
| 4063671 | |
| ≤80% | |
| Odorless | |
| Sodium bismuth oxide |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 235-455-6
RTECSNumber : VZ1750000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only