Learn More
Sodium bisulfate, 92%, Tech., granular, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 214850010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium bisulfate | |
7681-38-1 | |
Granules | |
92% | |
NaHSO4 | |
15, 8722 | |
Solubility in water: soluble. Other solubilities: decomposes by alcohol into na2so4 and h2so4 | |
[Na+].OS([O-])(=O)=O | |
120.05 | |
120.06 | |
Technical | |
2.1 |
177.0°C to 180.0°C | |
White to Yellow | |
1 kg | |
HNaO4S | |
MFCD00003531 | |
sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect | |
WBHQBSYUUJJSRZ-UHFFFAOYSA-M | |
sodium hydrogen sulfate | |
516919 | |
92% | |
Plastic Bottle | |
2.1000g/mL |
Chemical Identifiers
7681-38-1 | |
120.05 | |
WBHQBSYUUJJSRZ-UHFFFAOYSA-M | |
516919 | |
[Na+].OS([O-])(=O)=O |
HNaO4S | |
MFCD00003531 | |
sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect | |
sodium hydrogen sulfate |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
EINECSNumber : 231-665-7
RUO – Research Use Only