missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
![](http://beta-static.fishersci.ca/images/White.gif)
Sodium Bisulfate Monohydrate (Crystalline/Certified), Fisher Chemical™
Supplier: Thermo Fisher Scientific S240500
Specifications
Sodium Bisulfate Monohydrate | |
White | |
1 | |
500 g | |
MFCD00149210 | |
JXHZRQHZVYDRGX-UHFFFAOYSA-M | |
sodium hydrate hydrogen sulfate | |
23673662 | |
Certified | |
Poly Bottle | |
2.12g/cm³ |
58°C | |
10034-88-5 | |
Solid | |
H3NaO5S | |
sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox | |
O.[Na+].OS([O-])(=O)=O | |
138.07 | |
138.07 | |
Pass Test | |
430°C |
Chemical Identifiers
10034-88-5 | |
138.07 | |
JXHZRQHZVYDRGX-UHFFFAOYSA-M | |
23673662 | |
O.[Na+].OS([O-])(=O)=O |
H3NaO5S | |
MFCD00149210 | |
sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox | |
sodium hydrate hydrogen sulfate |
Safety and Handling
![](http://beta-static.fishersci.ca/images/White.gif)
DOTInformation : DOT Class 8, : Corrosive