missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium bisulfite, ACS reagent, powder, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 419440010

 View more versions of this product

Catalog No. AC419440010

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Specifications

Sodium bisulfite
7631-90-5
Crystalline Powder
NaO3S
MFCD00003530
15, 8724
Solubility in water: 300g/L. Other solubilities: soluble in 2 parts boiling water, soluble in about 70 parts alcohol
[Na+].[O-]S([O-])=O
103.05
CHEBI:26709
≥58.5% (SO2)
Plastic bottle
1.4800g/mL
150.0°C
White
1 kg
NaHSO3
01,1047; 11,487; 16,272
sodium bisulfite, sodium hydrogen sulfite, sodium bisulphite, sodium hydrogensulfite, sodium sulfhydrate, monosodium sulfite, uantax sbs, sulfurous acid, monosodium salt, caswell no. 750, hydrogen sodium sulfite
DWAQJAXMDSEUJJ-UHFFFAOYSA-L
sodium;hydrogen sulfite
23665763
104.06
ACS Reagent
1.48

Chemical Identifiers

7631-90-5
103.05
DWAQJAXMDSEUJJ-UHFFFAOYSA-L
23665763
sodium;hydrogen sulfite
NaO3S
MFCD00003530
sodium bisulfite, sodium hydrogen sulfite, sodium bisulphite, sodium hydrogensulfite, sodium sulfhydrate, monosodium sulfite, uantax sbs, sulfurous acid, monosodium salt, caswell no. 750, hydrogen sodium sulfite
CHEBI:26709
[Na+].[O-]S([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Contact with acids liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 231-548-0

RTECSNumber : VZ2000000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only