missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borate, 40 g/L, Ricca Chemical

Supplier:  Ricca Chemical Company 7852416

Catalog No. 7852416

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Borate
7732-18-5,1303-96-4
98.0,3.93
500 mL
Laboratory
Natural Poly Bottle
Colorless
94.0,3.77
Liquid
381.37 g/mol
40g/L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions