missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium borodeuteride, 99% (Isotopic), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03510206

 View more versions of this product

Catalog No. AA3510206


Explore available promotions

Description

Description

Sodium borodeuteride is used to prepare alfa, alfa-dideutero-beta-amino acids. It is also used to prepare amine-stabilized, deuterated borane-tetrhydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Powder
Moisture
White
15681-89-7
∼400°C (decomposition)
NaBD4
UN1426
Soluble in liquid ammonia,amines and pyridine.
[BH4-].[Na+]
37.83
41.88
5 g
Sodium borodeuteride
Isotopic
11
BH4Na
MFCD00003519
sodium borodeuteride, sodium tetradeuterioboron, sodium borohydride-d4, sodium 2h4 tetrahydroborate 1-, sulfatase, borate 1-, tetrahydro-d4-, sodium, nabd4, sodium 2h4 boranuide, deuterated sodium borohydride
YOQDYZUWIQVZSF-UHFFFAOYSA-N
sodium boranuide
23673181
99%

Chemical Identifiers

15681-89-7
37.83
YOQDYZUWIQVZSF-UHFFFAOYSA-N
23673181
[BH4-].[Na+]
BH4Na
MFCD00003519
sodium borodeuteride, sodium tetradeuterioboron, sodium borohydride-d4, sodium 2h4 tetrahydroborate 1-, sulfatase, borate 1-, tetrahydro-d4-, sodium, nabd4, sodium 2h4 boranuide, deuterated sodium borohydride
sodium boranuide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H330-H314-H318-H261
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
In contact with water releases flammable gas.

P223-P231+P232-P260-P264b-P270-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P378q-P380-P501c

H261-H301-H314

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: SODIUM BOROHYDRIDE

EINECSNumber : 239-764-7

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only