missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Borohydride (Powder), Fisher Chemical™

Supplier:  Thermo Fisher Scientific S67810

 View more versions of this product

Catalog No. S678-10



Explore available promotions

Description

Description

Specifications

Specifications

Sodium Borohydride
White
11
10 g
MFCD00003518
[BH4-].[Na+]
37.83
Pass Test
Negligible
360°C
16940-66-2
Solid
BH4Na
YOQDYZUWIQVZSF-UHFFFAOYSA-N
sodium boranuide
37.83
Poly Bottle
>220°C

Chemical Identifiers

16940-66-2
37.83
YOQDYZUWIQVZSF-UHFFFAOYSA-N
[BH4-].[Na+]
BH4Na
MFCD00003518
sodium boranuide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Water reactive. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Hygroscopic. Use personal protective equipment. Do not allow contact with water. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:1
Instability:2

DOTInformation : DOT Class 4.3, : Dangerous When Wet

SDS Documents

Documents

Promotions

Promotions