missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bromide (Granular/Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical S2553
Specifications
| 1390°C | |
| 755°C | |
| 7647-15-6 | |
| Powder | |
| ≥99 % | |
| MFCD00003475 | |
| JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
| sodium bromide | |
| 253881 | |
| 102.89 | |
| Certified ACS | |
| Glass Bottle |
| Sodium Bromide | |
| White | |
| 5 to 8.8 | |
| 3 kg | |
| BrNa | |
| sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
| [Na+].[Br-] | |
| 102.89 | |
| CHEBI:63004 | |
| ≥99% | |
| Pass Test |
Chemical Identifiers
| 7647-15-6 | |
| 102.89 | |
| JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
| 253881 | |
| sodium bromide |
| BrNa | |
| MFCD00003475 | |
| sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
| CHEBI:63004 | |
| [Na+].[Br-] |
Safety and Handling