missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium caprylate, 98%, Thermo Scientific Chemicals

$155.76 - $1782.58

Chemical Identifiers

CAS 1984-06-1
Molecular Formula C8H15NaO2
Molecular Weight (g/mol) 166.2
MDL Number MFCD00058511
InChI Key BYKRNSHANADUFY-UHFFFAOYSA-M
Synonym sodium octanoate, sodium caprylate, octanoic acid, sodium salt, sodium n-octanoate, sodium octoate, natrium octanoat, caprylic acid sodium salt, octanoic acid sodium salt, octanoic acid, sodium salt 1:1, unii-9xtm81vk2b
PubChem CID 23664772
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC334601000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
334601000
100 g
Each for $155.76
 
AC334600025
SDS View Documents
Thermo Scientific Chemicals
334600025
2.5 kg
Each for $1,782.58
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1984-06-1
166.2
BYKRNSHANADUFY-UHFFFAOYSA-M
23664772
C8H15NaO2
MFCD00058511
sodium octanoate, sodium caprylate, octanoic acid, sodium salt, sodium n-octanoate, sodium octoate, natrium octanoat, caprylic acid sodium salt, octanoic acid sodium salt, octanoic acid, sodium salt 1:1, unii-9xtm81vk2b

Specifications

1984-06-1
White
Plastic bottle
MFCD00058511
sodium octanoate, sodium caprylate, octanoic acid, sodium salt, sodium n-octanoate, sodium octoate, natrium octanoat, caprylic acid sodium salt, octanoic acid sodium salt, octanoic acid, sodium salt 1:1, unii-9xtm81vk2b
BYKRNSHANADUFY-UHFFFAOYSA-M
23664772
98%
Sodium caprylate, 98%
>225°C
Authentic
C8H15NaO2
100 g
Solubility in water: soluble in water
166.2
166.2
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-850-5

RTECSNumber : RH0787000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only