missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate, 99.999%, (trace metal basis)

CAS: 497-19-8 | CNa2O3 | 105.99 g/mol

$122.24 - $409.29

Identifiants chimiques

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O
voir les spécifications

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Qty  
Catalog Number Mfr. No. Quantity Packaging Price Qty  
AC436800050
SDS View Documents
Thermo Scientific Chemicals
436800050
5 g Glass bottle
chaque for $122.24
Only null left
 
AC436800250
SDS View Documents
Thermo Scientific Chemicals
436800250
25 g Glass bottle
chaque for $409.29
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

497-19-8
105.99
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
[Na+].[Na+].[O-]C([O-])=O
CNa2O3
MFCD00003494
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
CHEBI:29377

Specifications

1600.0°C
497-19-8
Crystalline Powder
99.999%
Na2CO3
15,8731
Solubility in water: 22 g/100mL (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
[Na+].[Na+].[O-]C([O-])=O
105.99
CHEBI:29377
99.999%
Glass bottle
851.0°C
White
5 g
CNa2O3
MFCD00003494
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium carbonate
10340
105.99
Trace Metal Basis
Sodium carbonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed or concerned: Get med

GHS Signal Word: Warning

EINECSNumber : 207-838-8

SDS
Documents

Documents

RUO – Research Use Only