Learn More
Sodium carbonate decahydrate, 99+%
CAS: 6132-02-1 | CH20Na2O13 | 286.14 g/mol
$78.19 - $840.29
Chemical Identifiers
| CAS | 2-1-6132 |
|---|---|
| Molecular Formula | CH20Na2O13 |
| Molecular Weight (g/mol) | 286.14 |
| MDL Number | MFCD00149178 |
| InChI Key | XYQRXRFVKUPBQN-UHFFFAOYSA-L |
| Synonym | sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t |
| PubChem CID | 151402 |
| IUPAC Name | disodium;carbonate;decahydrate |
| SMILES | C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1748436
|
Thermo Scientific Chemicals
A1748436 |
500 g |
Each for $78.19
|
|
|||||
|
AAA174840E
|
Thermo Scientific Chemicals
A174840E |
2500 g |
Each for $259.49
|
|
|||||
|
AAA174840C
|
Thermo Scientific Chemicals
A174840C |
10,000 g |
Each for $840.29
|
|
|||||
Description
Sodium carbonate decahydrate is used as a raw material in various industries like glass, paper making, soap, detergent, textile and leather. It is also used as a food additive. It is also involved as a precipitation agent of carbonate as well as an analytical reagent in laboratories. It plays an important role as strong base and powerful electrolyte. It finds application in metal refining and cement production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium carbonate decahydrate is used as a raw material in various industries like glass, paper making, soap, detergent, textile and leather. It is also used as a food additive. It is also involved as a precipitation agent of carbonate as well as an analytical reagent in laboratories. It plays an important role as strong base and powerful electrolyte. It finds application in metal refining and cement production.
Solubility
Soluble in water and glycerol. Insoluble in alcohol.
Notes
Air sensitive. Incompatible with strong acids and aluminum. Keep the container tightly closed.
Chemical Identifiers
| 2-1-6132 | |
| 286.14 | |
| XYQRXRFVKUPBQN-UHFFFAOYSA-L | |
| 151402 | |
| C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+] |
| CH20Na2O13 | |
| MFCD00149178 | |
| sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t | |
| disodium;carbonate;decahydrate |
Specifications
| 32°C to 34°C | |
| 500 g | |
| Na2CO3·10H2O | |
| 14,8596 | |
| Soluble in water and glycerol. Insoluble in alcohol. | |
| C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+] | |
| 286.14 | |
| 286.14 (105.99 Anhydrous) | |
| 1.46 |
| 2-1-6132 | |
| CH20Na2O13 | |
| MFCD00149178 | |
| sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t | |
| XYQRXRFVKUPBQN-UHFFFAOYSA-L | |
| disodium;carbonate;decahydrate | |
| 151402 | |
| ≥99% | |
| Sodium carbonate decahydrate |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 207-838-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only