missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium carbonate monohydrate, 99.6%, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 419480051

 View more versions of this product

Catalog No. AC419480051


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium carbonate monohydrate
5968-11-6
Crystalline Powder
99.6%
Na2CO3·H2O
07,322
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
MQRJBSHKWOFOGF-UHFFFAOYSA-L
disodium;carbonate;hydrate
2735133
99.6%
13.0% to 15.0% (150°C)
851.0°C
White
5 kg
CNa2O3·H2O
MFCD00149177
15, 8731
Solubility in water: 210g/L. Other solubilities: soluble in 1.8 parts boiling water,in 7 parts glyceral,insoluble in alcohol
C(=O)([O-])[O-].O.[Na+].[Na+]
124.01
124.01
ACS Reagent
Plastic bottle

Chemical Identifiers

5968-11-6
124.01
MQRJBSHKWOFOGF-UHFFFAOYSA-L
2735133
C(=O)([O-])[O-].O.[Na+].[Na+]
CNa2O3·H2O
MFCD00149177
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
disodium;carbonate;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only