missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate monohydrate, ACS, 99.5% min

CAS: 5968-11-6 | CH2Na2O4 | 124.00 g/mol

$70.97 - $264.79

Chemical Identifiers

CAS 5968-11-6
Molecular Formula CH2Na2O4
Molecular Weight (g/mol) 124.00
MDL Number MFCD00149177
InChI Key MQRJBSHKWOFOGF-UHFFFAOYSA-L
Synonym sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
PubChem CID 2735133
IUPAC Name disodium;carbonate;hydrate
SMILES C(=O)([O-])[O-].O.[Na+].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3648522
View Documents
Thermo Scientific Chemicals
03648522
100 g
Each for $70.97
Only null left
 
AA3648536
View Documents
Thermo Scientific Chemicals
03648536
500 g
Each for $129.75
Only null left
 
AA36485A3
View Documents
Thermo Scientific Chemicals
036485A3
2 kg
Each for $264.79
Only null left
 
Description

Description

It is widely employed as a water softener, an electrolyte, as a good conductor in electrolysis, as a primary standard for acid-base titrations and mostly as a buffer component in applications such as chromatography, capillary electrophoresis and enzyme catalysis. Also used in glass manufacture, detergent manufacture, sodium chemicals, carbonate chemicals manufacture, pulp and paper, brine treatment, flue gas desulfurization, coal treatment and ion exchange resin regeneration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5968-11-6
124.00
MQRJBSHKWOFOGF-UHFFFAOYSA-L
2735133
C(=O)([O-])[O-].O.[Na+].[Na+]
CH2Na2O4
MFCD00149177
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
disodium;carbonate;hydrate

Specifications

100°C-H2O
Granules
99.5%
MFCD00149177
Soluble in water and glycerol. Insoluble in alcohol,acetone,alkyl acetates,benzonitrile and liquid ammonia.
C(=O)([O-])[O-].O.[Na+].[Na+]
124.00
124.01 (105.99 Anhydrous)
Hygroscopic
Sodium carbonate monohydrate, For analysis ACS
5968-11-6
100 g
CH2Na2O4
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
MQRJBSHKWOFOGF-UHFFFAOYSA-L
disodium;carbonate;hydrate
2735133
ACS Reagent
2.250 g/mL
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H336

EINECSNumber : 207-838-8

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only