missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Chloride 0.9% (Normal Saline), USP, Sterile Grade, Cytiva Hyclone™

Sodium Chloride Injection, USP is a sterile solution of sodium chloride in water for injection. It contains no antimicrobial agents. It contains NLT 95.0% and NMT 105.0% of the labeled amount of sodium chloride (NaCl).

Supplier:  Cytiva Z1379

 View more versions of this product

Catalog No. Z1379


Explore available promotions

Description

Description

  • Manufactured under cGMP and ISO 13485 guidelines in a FDA registered facility
  • Sterile and endotoxin tested
  • Ideal for medical device and biopharmaceutical manufacturing
  • Suitable in research, testing, cell culture and molecular protocols
Specifications

Specifications

20 L
MFCD00003477
[Na+].[Cl-]
58.44
Bioprocess Bag
ClNa
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
0.009

Chemical Identifiers

ClNa
MFCD00003477
sodium chloride
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
[Na+].[Cl-]
Videos
SDS
Documents

Documents

Promotions

Promotions