missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Sodium Chloride (Granular/USP/FCC), Fisher Chemical™
Supplier: Fisher Chemical S640500
Specifications
1461°C | |
801°C | |
White | |
Solid | |
ClNa | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
99.0 to 100.5% (FCC) | |
Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC) | |
Poly Bottle | |
2.165g/cm³ |
Sodium Chloride | |
7647-14-5 | |
5.0 to 8.0 | |
500 g | |
MFCD00003477 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
5234 | |
58.44 | |
FCC/USP | |
0.5% max. (FCC), 0.5% max. (USP), 0.5% max. (USP/FCC) | |
1mmHg at 440°C |
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
5234 | |
sodium chloride |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
CHEBI:26710 | |
[Na+].[Cl-] |
Safety and Handling

CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:0
Instability:1