missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Chloride, Powder, USP, 99-100.5%, Spectrum™ Chemical
SO160, 7647-14-5, NaCl
Supplier: Spectrum Chemical Mfg Cor SO16050KGBL
Description
Spectrum™ Chemical Sodium Chloride, Powder, USP is used in the formulation of oral hygiene products, shampoos, fragrance, skin, hair, nail, cleansing, suntan, makeup and bath products. This grade is specifically used in oral care applications. This product is not sterile and not intended for use in the manufacture of injectable dosage forms, peritoneal dialysis solutions, hemodialysis solutions or hemofiltration solutions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
7647-14-5 | |
50 kg | |
MFCD00003477 | |
[Na+].[Cl-] | |
58.44 | |
USP | |
Fiber Drum |
1 | |
ClNa | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
99 to 100.5% | |
0.005 |
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
[Na+].[Cl-] |
ClNa | |
MFCD00003477 | |
sodium chloride |