missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Chloride Standard, 1000 ppm NaCl, Ricca Chemical
Supplier: Ricca Chemical Company R72231001A
Specifications
Sodium Chloride Standard | |
7647-14-5,7732-18-5 | |
1 L | |
ClNa | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
5234 | |
Odorless |
1000 ppm NaCl | |
Natural Poly Bottle | |
Colorless | |
MFCD00003477 | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
Liquid |
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
CHEBI:26710 | |
[Na+].[Cl-] |
ClNa | |
MFCD00003477 | |
5234 | |
sodium chloride |
Safety and Handling
ShelfLife : 18 months