missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium chlorite, 80%, pure, unstabilized, Thermo Scientific Chemicals

Sodium chlorite, 80%, ClNaO2, CAS Number-7758-19-2, 7647-14-5, unii-g538ebv4vf, alcide ld, neo silox d, chlorous acid, sodium salt, caswell no. 755, sodiumchlorite, textone, sodium chlorite, textile, chlorite sodium, 500g, CHEBI:78667, White, 231-836-6, 05,603; 09,423; 11,481; 13,280, 90.44, Pure

Supplier:  Thermo Scientific Chemicals 223230050

 View more versions of this product

Catalog No. AC223230050

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium chlorite
Unstabilized
White
5 g
ClNaO2
MFCD00003478
15, 8735
Solubility in water: 390g/L (17°C).
[O-]Cl=O.[Na+]
90.44
CHEBI:78667
80%
Glass Bottle
180.0°C to 200.0°C
7758-19-2,7647-14-5
Powder
75% min. (Iodometry)
NaClO2
05,603; 09,423; 11,481; 13,280
sodium chlorite, chlorous acid, sodium salt, textone, chlorite sodium, textile, alcide ld, neo silox d, caswell no. 755, sodiumchlorite, unii-g538ebv4vf
UKLNMMHNWFDKNT-UHFFFAOYSA-M
sodium;chlorite
23668197
90.44
Pure

Chemical Identifiers

7758-19-2
UKLNMMHNWFDKNT-UHFFFAOYSA-M
23668197
sodium;chlorite
MFCD00003478
sodium chlorite, chlorous acid, sodium salt, textone, chlorite sodium, textile, alcide ld, neo silox d, caswell no. 755, sodiumchlorite, unii-g538ebv4vf
CHEBI:78667
[O-]Cl=O.[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
May cause fire or explosion; strong oxidizer.
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cauti

GHS Signal Word: Danger

EINECSNumber : 231-836-6

RTECSNumber : VZ4800000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only