missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium chromate tetrahydrate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1749936

 View more versions of this product

Catalog No. AAA1749936


Explore available promotions

Description

Description

Sodium chromate tetrahydrate is used as a corrosion inhibitor, wood preservative and an auxiliary in textile industry. It serves as an oxidant, which is used to prepare carboxylic acids and ketones from primary and secondary alcohols respectively. Further, it is used as a starting material to prepare other chromium compounds and as a catalyst in organic synthesis especially pharmaceuticals and agro chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium chromate tetrahydrate is used as a corrosion inhibitor, wood preservative and an auxiliary in textile industry. It serves as an oxidant, which is used to prepare carboxylic acids and ketones from primary and secondary alcohols respectively. Further, it is used as a starting material to prepare other chromium compounds and as a catalyst in organic synthesis especially pharmaceuticals and agro chemicals.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible withstrong reducing agents, organic materials, strong acids, combustible materials and powdered metals.
Specifications

Specifications

Sodium chromate tetrahydrate
500 g
MFCD00149167
14,8601
Soluble in water.
O.O.O.O.[O-][Cr](=O)(=O)[O-].[Na+].[Na+]
234.032
234.04 (161.97 Anhydrous)
Odorless
10034-82-9
CrH8Na2O8
UN3288
sodium chromate tetrahydrate, cro4.2na.4h2o, acmc-20ajol, disodium chromate tetrahydrate, ksc492q3d, disodium chromate 2-tetrahydrate, sodium chromate tetrahydrate, reagent grade, sodium chromate tetrahydrate, p.a
AVHUVTPOXZEIRG-UHFFFAOYSA-N
disodium;dioxido(dioxo)chromium;tetrahydrate
22146515
≥99%
2.73

Chemical Identifiers

10034-82-9
234.032
AVHUVTPOXZEIRG-UHFFFAOYSA-N
22146515
O.O.O.O.[O-][Cr](=O)(=O)[O-].[Na+].[Na+]
CrH8Na2O8
MFCD00149167
sodium chromate tetrahydrate, cro4.2na.4h2o, acmc-20ajol, disodium chromate tetrahydrate, ksc492q3d, disodium chromate 2-tetrahydrate, sodium chromate tetrahydrate, reagent grade, sodium chromate tetrahydrate, p.a
disodium;dioxido(dioxo)chromium;tetrahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H334-H340-H350-H360-H372-H314-H318-H312-H317
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H301-H312-H314-H317-H330-H334-H335-H340-H350-H360-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-889-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only