missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Citrate, Anhydrous, USP, 99-100.5%, Spectrum™ Chemical
S1986, 68-04-2, C6H5Na3O7
$704.21 - $7570.91
Chemical Identifiers
CAS | 68-04-2 |
---|---|
Molecular Formula | C6H5Na3O7 |
Molecular Weight (g/mol) | 258.07 |
MDL Number | MFCD00012462 |
InChI Key | HRXKRNGNAMMEHJ-UHFFFAOYSA-K |
IUPAC Name | trisodium 2-hydroxypropane-1,2,3-tricarboxylate |
SMILES | [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18606449
|
Spectrum Chemical Mfg Cor
S1986500GM |
500 g | Poly Bottle |
Each for $704.21
|
|
||||
18606450
|
Spectrum Chemical Mfg Cor
S198625KG |
2.5 kg | Poly Pail |
Each for $1,873.79
|
|
||||
18606451
|
Spectrum Chemical Mfg Cor
S198612KG |
12 kg | Fiber Drum |
Each for $3,868.41
|
|
||||
18606453
|
Spectrum Chemical Mfg Cor
S198612KGPL |
12 kg | Poly Pail |
Each for $3,907.15
|
|
||||
18606452
|
Spectrum Chemical Mfg Cor
S198645KGBL |
45 kg | Fiber Drum |
Each for $7,570.91
|
|
||||
Description
Spectrum™ Chemical Sodium Citrate, Anhydrous, USP is used to treat certain metabolic problems (acidosis) caused by kidney disease. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
68-04-2 | |
258.07 | |
HRXKRNGNAMMEHJ-UHFFFAOYSA-K | |
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O |
C6H5Na3O7 | |
MFCD00012462 | |
trisodium 2-hydroxypropane-1,2,3-tricarboxylate |
Specifications
68-04-2 | |
Poly Bottle | |
MFCD00012462 | |
HRXKRNGNAMMEHJ-UHFFFAOYSA-K | |
trisodium 2-hydroxypropane-1,2,3-tricarboxylate | |
99 to 100.5% |
1 | |
C6H5Na3O7 | |
500 g | |
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O | |
258.07 | |
USP |