missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate Dihydrate (Granular/Certified), Fisher Chemical™

$271.42 - $8806.33

Chemical Identifiers

CAS 4-3-6132
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
MDL Number MFCD00150031
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
S279-500
SDS View Documents Promotion Details
Thermo Fisher Scientific
S279500
500 g Poly Bottle
Each for $271.42
Case of 6 Each for $1,628.52
Only null left
 
S279-3
SDS View Documents Promotion Details
Fisher Chemical
S2793
3 kg Poly Bottle
Each for $705.00
Case of 4 Each for $2,820.00
Only null left
 
S279-10
SDS View Documents Promotion Details
Fisher Chemical
S27910
10 kg Poly Pail
Each for $1,900.00
Only null left
 
S279-50
SDS Accessories View Documents Promotion Details
Thermo Fisher Scientific
FLS27950
50 kg Fiber Drum
Each for $8,806.33
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4-3-6132
294.10
NLJMYIDDQXHKNR-UHFFFAOYSA-K
71474
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
C6H9Na3O9
MFCD00150031
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
CHEBI:32142
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

4-3-6132
300°C
1.67g/mL
Poly Bottle
MFCD00150031
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
NLJMYIDDQXHKNR-UHFFFAOYSA-K
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
71474
294.1
Sodium Citrate Dihydrate
>230°C
White
7.5 to 9.5
C6H9Na3O9
500 g
0.005% max. Insoluble Matter
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
294.10
CHEBI:32142
Solid
Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

SDS
missing translation for 'documents'

missing translation for 'documents'