Learn More
Sodium cyanide, 98+%, extra pure, Thermo Scientific Chemicals
$92.49 - $281.46
Chemical Identifiers
CAS | 143-33-9 |
---|---|
Molecular Formula | CNNa |
Molecular Weight (g/mol) | 49.01 |
MDL Number | MFCD00003523 |
InChI Key | MNWBNISUBARLIT-UHFFFAOYSA-N |
Synonym | sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h |
PubChem CID | 8929 |
SMILES | [Na+].[C-]#N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC370310050
|
Thermo Scientific Chemicals
370310050 |
5 g | Glass bottle |
Each for $92.49
|
|
||||
AC370310010
|
Thermo Scientific Chemicals
370310010 |
1 kg | Glass bottle |
Each for $281.46
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
143-33-9 | |
49.01 | |
MNWBNISUBARLIT-UHFFFAOYSA-N | |
8929 |
CNNa | |
MFCD00003523 | |
sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h | |
[Na+].[C-]#N |
Specifications
1497.0°C | |
143-33-9 | |
Crystals | |
98% min. (Argentometry) | |
NaCN | |
02, IV, 50 | |
15, 8741 | |
Solubility in water: 370g/L (20°C). Other solubilities: sparingly soluble in ethanol | |
[Na+].[C-]#N | |
49.01 | |
49 | |
Extra Pure | |
Sodium cyanide, 98+% |
562.0°C | |
White | |
5 g | |
CNNa | |
MFCD00003523 | |
04,446; 05,506; 06,535; 07,333; 08,453; 11,481; 13,185 | |
sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h | |
MNWBNISUBARLIT-UHFFFAOYSA-N | |
sodium;cyanide | |
8929 | |
≥98% | |
Glass bottle |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lastin
GHS P Statement
Absorb spillage to prevent material damage.
Wear protective gloves/protective clothing.
Rinse mouth.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a p
GHS Signal Word: Danger
EINECSNumber : 205-599-4
Recommended Storage : Poison Room
RUO – Research Use Only