Learn More
Sodium cyanoborohydride, 95%, Thermo Scientific Chemicals
Selective Reducing Agent For Aldehydes, Ketones, Oximes
$57.59 - $708.95
Chemical Identifiers
CAS | 25895-60-7 |
---|---|
Molecular Formula | CH3BNNa |
Molecular Weight (g/mol) | 62.84 |
MDL Number | MFCD00003516 |
InChI Key | CVDUGUOQTVTBJH-UHFFFAOYSA-N |
Synonym | sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1- |
PubChem CID | 20587905 |
SMILES | [Na+].[BH3-]C#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA8783906
|
Thermo Scientific Chemicals
08783906 |
5 g |
Each for $57.59
|
|
|||||
AA8783914
|
Thermo Scientific Chemicals
08783914 |
25 g |
Each for $190.95
|
|
|||||
AA8783922
|
Thermo Scientific Chemicals
08783922 |
100 g |
Each for $708.95
|
|
|||||
Description
Selective reducing agent for aldehydes, ketones, oximes, enamines. Reductive animation of ketones, aldehydes. Reductive alkylation of amines, hydrazines Sodium cyanoborohydride is used as a reagent involved in the reductive amination of aldehydes, ketones, oximes and enamines. It is also used in the reductive alkylation of amines and hydrazines. It acts as a catalyst in the preparation of phenolate-bridged dilanthanum(III) complex. In addition, it is used in the conversion of imines to amines. It plays an important role in the reductive deoxygenation of ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Moisture Sensitive; Highly flammableChemical Identifiers
25895-60-7 | |
62.84 | |
CVDUGUOQTVTBJH-UHFFFAOYSA-N | |
20587905 |
CH3BNNa | |
MFCD00003516 | |
sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1- | |
[Na+].[BH3-]C#N |
Specifications
>300°C (decomposition) | |
White | |
5 g | |
CH3BNNa | |
sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1- | |
CVDUGUOQTVTBJH-UHFFFAOYSA-N | |
sodium;cyanoboron(1-) | |
20587905 | |
Moisture sensitive | |
Sodium cyanoborohydride |
25895-60-7 | |
Powder | |
95% | |
MFCD00003516 | |
Soluble in water,THF,methanolSoluble in water,diglyme,tetrahydrofuran and methanol. Slightly soluble in dimethyl sulfoxide. Insoluble in diethyl ether. | |
[Na+].[BH3-]C#N | |
62.84 | |
62.84 | |
1.20 g/cm3 at 20°C |
Safety and Handling
P210-P223-P231+P232-P240-P241-P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H228-H261-H300+H310+H330-H314
DOTInformation : DOT Class: 4.3, Packaging Group: II
EINECSNumber : 247-317-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only