missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Cyanoborohydride 95.0+%, TCI America™

Supplier:  TCI America S03965G

 View more versions of this product

Catalog No. S03965G


Explore available promotions

Description

Description

Specifications

Specifications

Sodium Cyanoborohydride [Reducing Agent]
25895-60-7
Crystalline Powder
CH3BNNa
3134
CVDUGUOQTVTBJH-UHFFFAOYSA-N
sodium cyanoboranuide
20587905
≥95.0% (T)
White-Yellow
8 to 9 (10% H2O soln.)
5 g
MFCD00003516
sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
[Na+].[BH3-]C#N
62.84
62.84

Chemical Identifiers

25895-60-7
62.84
CVDUGUOQTVTBJH-UHFFFAOYSA-N
20587905
[Na+].[BH3-]C#N
CH3BNNa
MFCD00003516
sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
sodium cyanoboranuide
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions