Learn More
Sodium cyclopentadienide, 2-3M in THF, Thermo Scientific Chemicals
Reagent for the preparation of metallocenes
$166.01 - $664.73
Chemical Identifiers
CAS | 4984-82-1 |
---|---|
Molecular Formula | C5H5Na |
Molecular Weight (g/mol) | 88.085 |
MDL Number | MFCD00012161 |
InChI Key | JQUZTGJSSQCTPV-UHFFFAOYSA-N |
Synonym | cyclopentadienylsodium, sodium cyclopenta-1,3-dien-1-ide, sodium cyclopentadienide, 2.0m solution in tetrahydrofuran |
PubChem CID | 21915903 |
IUPAC Name | sodium;cyclopenta-1,3-diene |
SMILES | C1C=CC=[C-]1.[Na+] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4984-82-1 | |
88.085 | |
JQUZTGJSSQCTPV-UHFFFAOYSA-N | |
21915903 | |
C1C=CC=[C-]1.[Na+] |
C5H5Na | |
MFCD00012161 | |
cyclopentadienylsodium, sodium cyclopenta-1,3-dien-1-ide, sodium cyclopentadienide, 2.0m solution in tetrahydrofuran | |
sodium;cyclopenta-1,3-diene |
Specifications
4984-82-1 | |
0.938 g/mL | |
Ether-like | |
MFCD00012161 | |
UN3399 | |
cyclopentadienylsodium, sodium cyclopenta-1,3-dien-1-ide, sodium cyclopentadienide, 2.0m solution in tetrahydrofuran | |
JQUZTGJSSQCTPV-UHFFFAOYSA-N | |
sodium;cyclopenta-1,3-diene | |
21915903 | |
2 to 3M in THF | |
Sodium cyclopentadienide |
Pink to Red | |
−17°C (1°F) | |
C5H5Na | |
0.1 mol | |
Air and moisture sensitive | |
Reacts with water. | |
C1C=CC=[C-]1.[Na+] | |
88.085 | |
88.09 | |
Liquid |
Safety and Handling
GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H225-H250-H261-H302-H314-H335-H336-H351
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
EINECSNumber : 225-636-8
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only