Learn More
Sodium D-gluconate, 97%, Thermo Scientific Chemicals
$183.03 - $636.29
Chemical Identifiers
CAS | 527-07-1 |
---|---|
Molecular Formula | C6H11NaO7 |
Molecular Weight (g/mol) | 218.137 |
MDL Number | MFCD00064210 |
InChI Key | UPMFZISCCZSDND-JJKGCWMISA-M |
Synonym | sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100t |
PubChem CID | 23672301 |
ChEBI | CHEBI:84997 |
IUPAC Name | sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate |
SMILES | C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA104640E
|
Thermo Scientific Chemicals
A104640E |
2500 g |
Each for $183.03
|
|
|||||
AAA104640C
|
Thermo Scientific Chemicals
A104640C |
10 kg |
Each for $636.29
|
|
|||||
Description
Sodium D-gluconate is a good chelator at alkaline pH, with chelating power relatively better than EDTA, NTA and other chelators. It has wide range of industrial applications which include textile dyeing, water treatments, printing and as cleaning agent for glass bottles. It also finds use as derusting agent in metallurgy and as additive in cement. It is used in food stuffs as bitterness inhibitor and as electrolyte replenisher.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium D-gluconate is a good chelator at alkaline pH, with chelating power relatively better than EDTA, NTA and other chelators. It has wide range of industrial applications which include textile dyeing, water treatments, printing and as cleaning agent for glass bottles. It also finds use as derusting agent in metallurgy and as additive in cement. It is used in food stuffs as bitterness inhibitor and as electrolyte replenisher.
Solubility
Very soluble in water; sparingly soluble in alcohol; insoluble in ether.
Notes
In general it is stable, non corrosive, non-toxic, readily biodegradable and incompatible with strong oxidizing agents.
Chemical Identifiers
527-07-1 | |
218.137 | |
UPMFZISCCZSDND-JJKGCWMISA-M | |
23672301 | |
sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate |
C6H11NaO7 | |
MFCD00064210 | |
sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100t | |
CHEBI:84997 | |
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+] |
Specifications
527-07-1 | |
C6H11NaO7 | |
2500 g | |
sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100t | |
UPMFZISCCZSDND-JJKGCWMISA-M | |
sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate | |
23672301 | |
218.14 |
∽206°C (decomposition) | |
MFCD00064210 | |
3919651 | |
Very soluble in water; sparingly soluble in alcohol; insoluble in ether. | |
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+] | |
218.137 | |
CHEBI:84997 | |
Sodium D-gluconate |
Safety and Handling
EINECSNumber : 208-407-7
RTECSNumber : LZ5235000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only