missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium dichromate dihydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 219240010

 View more versions of this product

Catalog No. AC219240010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

400.0°C
357.0°C
7789-12-0
3.5 to 3.9 (5% soln.)
1 kg
Cr2Na2O7·2H2O
MFCD00149166
15, 8744
Solubility in water: 730g/L (20°C). Other solubilities: insoluble in alcohol
O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]
298
298
Plastic Bottle
Sodium dichromate dihydrate
99%
Orange
Crystals or Needles
98.5% min. (Iodometry)
Na2Cr2O7·2H2O
01,1059; 03,265
sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
JYDRNIYTFCBIFC-UHFFFAOYSA-N
disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
108005
99%

Chemical Identifiers

7789-12-0
298
JYDRNIYTFCBIFC-UHFFFAOYSA-N
108005
O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]
Cr2Na2O7·2H2O
MFCD00149166
sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause genetic defects.
May cause an allergic skin reaction.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Har

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

RTECSNumber : HX7750000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only