missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium diethyldithiocarbamate trihydrate, 98%, Thermo Scientific Chemicals

$113.59 - $341.49

Chemical Identifiers

CAS 20624-25-3
Molecular Formula C5H16NNaO3S2
Molecular Weight (g/mol) 225.297
MDL Number MFCD00150617
InChI Key WWGXHTXOZKVJDN-UHFFFAOYSA-M
Synonym sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czech
PubChem CID 517546
IUPAC Name sodium;N,N-diethylcarbamodithioate;trihydrate
SMILES CCN(CC)C(=S)[S-].O.O.O.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1589822
View Documents
Thermo Scientific Chemicals
A1589822
100 g N/A
 
AAA1589836
View Documents
Thermo Scientific Chemicals
A1589836
500 g N/A
 
Description

Description

Sodium diethyldithiocarbamate trihydrate is a spin trapping reagent used in conjunction with ferrous ion to detect nitric oxide in brain, kidney, liver and other tissues. It is used as sensitive reagent for the determination of copper, rubber vulcanization accelerator and finds application in the determination of other metals such as zinc, cobalt, platinum and palladium. Furthermore, it is an inhibitor of superoxide dismutase, ascorbate oxidase and other enzymes as well as inhibits the induction of macrophage nitric oxide synthase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium diethyldithiocarbamate trihydrate is a spin trapping reagent used in conjunction with ferrous ion to detect nitric oxide in brain, kidney, liver and other tissues. It is used as sensitive reagent for the determination of copper, rubber vulcanization accelerator and finds application in the determination of other metals such as zinc, cobalt, platinum and palladium. Furthermore, it is an inhibitor of superoxide dismutase, ascorbate oxidase and other enzymes as well as inhibits the induction of macrophage nitric oxide synthase.

Solubility
Soluble in water, ethanol, methanol and acetone. Insoluble in ether.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

20624-25-3
225.297
WWGXHTXOZKVJDN-UHFFFAOYSA-M
517546
CCN(CC)C(=S)[S-].O.O.O.[Na+]
C5H16NNaO3S2
MFCD00150617
sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czech
sodium;N,N-diethylcarbamodithioate;trihydrate

Specifications

20624-25-3
C5H16NNaO3S2
100 g
Hygroscopic
sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czech
WWGXHTXOZKVJDN-UHFFFAOYSA-M
sodium;N,N-diethylcarbamodithioate;trihydrate
517546
98%
94°C to 102°C
MFCD00150617
3920507
14,3378
Soluble in water,ethanol,methanol and acetone. Insoluble in ether.
CCN(CC)C(=S)[S-].O.O.O.[Na+]
225.297
225.31 (171.28 Anhydrous)
Sodium diethyldithiocarbamate trihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 205-710-6

RTECSNumber : EZ6550000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only