missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium dihydrogen phosphate dihydrate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A113160E

 View more versions of this product

Catalog No. AAA113160E


Explore available promotions

Description

Description

Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates

Solubility
Soluble in water.(1 M at 20°C)

Notes
Hygroscopic. Store under inert gas. Incompatible with oxidizing agents and water.
Specifications

Specifications

Sodium dihydrogen phosphate dihydrate
2500 g
NaH2PO4·2H2O
14,8660
Soluble in water.(1 M at 20°C)
O.O.OP(=O)(O)[O-].[Na+]
156.006
156.01 (119.98 Anhydrous)
Hygroscopic
13472-35-0
H6NaO6P
MFCD00149209
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
VBJGJHBYWREJQD-UHFFFAOYSA-M
sodium;dihydrogen phosphate;dihydrate
23673460
99%
1.91

Chemical Identifiers

13472-35-0
MFCD00149209
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
sodium;dihydrogen phosphate;dihydrate
H6NaO6P
VBJGJHBYWREJQD-UHFFFAOYSA-M
23673460
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-449-2

RTECSNumber : WA1900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only