Learn More
Sodium dihydrogen phosphate dihydrate, 98+%
CAS: 13472-35-0 | H6NaO6P | 156.006 g/mol
Supplier: Thermo Scientific Chemicals A1131636
Description
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates
Solubility
Soluble in water.(1 M at 20°C)
Notes
Hygroscopic. Store under inert gas. Incompatible with oxidizing agents and water.
Specifications
| Sodium dihydrogen phosphate dihydrate | |
| 500 g | |
| NaH2PO4·2H2O | |
| 14,8660 | |
| Soluble in water.(1 M at 20°C) | |
| O.O.OP(=O)(O)[O-].[Na+] | |
| 156.006 | |
| 156.01 (119.98 Anhydrous) | |
| Hygroscopic |
| 13472-35-0 | |
| H6NaO6P | |
| MFCD00149209 | |
| sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
| VBJGJHBYWREJQD-UHFFFAOYSA-M | |
| sodium;dihydrogen phosphate;dihydrate | |
| 23673460 | |
| 99% | |
| 1.91 |
Chemical Identifiers
| 13472-35-0 | |
| MFCD00149209 | |
| sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
| sodium;dihydrogen phosphate;dihydrate |
| H6NaO6P | |
| VBJGJHBYWREJQD-UHFFFAOYSA-M | |
| 23673460 |
Safety and Handling
EINECSNumber : 231-449-2
RTECSNumber : WA1900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only