missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dihydrogen Phosphate Monohydrate
Supplier: Technidata 1063461000
Specifications
| Sodium Dihydrogen Phosphate Monohydrate | |
| 1 kg | |
| MFCD00149208 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| sodium phosphoric acid hydrate | |
| 516949 | |
| 99.0 to 102.0% | |
| Plastic Bottle |
| 10049-21-5 | |
| H5NaO5P | |
| sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
| O.[Na+].OP(O)(O)=O | |
| 139.00 | |
| 137.99 | |
| ACS | |
| 2.04 g/cm3 (20°C); Bulk Density: 880 kg/m3 |
Chemical Identifiers
| 10049-21-5 | |
| 139.00 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| 516949 | |
| O.[Na+].OP(O)(O)=O |
| H5NaO5P | |
| MFCD00149208 | |
| sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
| sodium phosphoric acid hydrate |
Safety and Handling
EINECSNumber : 231-449-2