missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium, dispersion, 40% in oil, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01034056

 View more versions of this product

Catalog No. AA1034056


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

White
7440-23-5
Na
4 x 250 g
KEAYESYHFKHZAL-UHFFFAOYSA-N
sodium
5360545
Air and moisture sensitive
Sodium
99+% (Metals basis)
MFCD00085307
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
[Na]
22.99
CHEBI:26708
Dispersion

Chemical Identifiers

7440-23-5
22.99
KEAYESYHFKHZAL-UHFFFAOYSA-N
5360545
sodium
Na
MFCD00085307
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
CHEBI:26708
[Na]
Videos
Safety and Handling

Safety and Handling

P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H260-H314-H335

DOTInformation : DOT Class: 4.3, Packaging Group: I

EINECSNumber : 231-132-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only