Learn More
![](http://beta-static.fishersci.ca/images/Red.gif)
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
For protein extraction procedures and SDS polyacrylamide gels
Supplier: Fisher BioReagents BP166100
Description
For protein extraction procedures and SDS polyacrylamide gels
- DNase and RNase-free detergent for protein extraction procedures and polyacrylamide gel electrophoresis
- SDS is a commonly used detergent in protein purification and polyacrylamide gel electrophoresis (PAGE)
- SDS denatures proteins by disrupting the non-covalent bonds of the protein
- Functionally tested for use in electrophoresis
- Intended for research use only; not for human or animal diagnostic purposes
Specifications
0.1 max. (0.1% solution) at 280nm, 0.4 max. (0.1% solution) at 230nm | |
White | |
Solid | |
DNase- and RNase-Free | |
(as Fatty Alcohol Sulfate) >=99% | |
CH3(CH2)10CH2OSO3Na | |
8.5 to 10 | |
15, 8768 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium dodecyl sulfate | |
Pass Test | |
CHEBI:8984 | |
≥99% (≥98% C12-OH) | |
Glass Bottle |
Sodium Dodecyl Sulfate | |
151-21-3,7647-14-5,7757-82-6 | |
206°C | |
(as Fatty Alcohol Sulfate) >=99% | |
C12H25NaO4S | |
MFCD00036175 | |
100 g | |
Sodium Lauryl Sulfate, SDS | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
3423265 | |
288.38 | |
Electrophoresis |
Chemical Identifiers
C12H25NaO4S | |
MFCD00036175 | |
Sodium Lauryl Sulfate, SDS | |
CHEBI:8984 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
3423265 | |
sodium dodecyl sulfate |
Safety and Handling
![](http://beta-static.fishersci.ca/images/Red.gif)
DANGER!
Emergency Overview
Flammable solid. Causes eye, skin, and respiratory tract irritation. May cause allergic respiratory reaction. Avoid contact with skin and eyes. Do not breathe dust. Use explosion-proof equipment. Use only non-sparking tools. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Remove from exposure, lie down. Move to fresh air.
NFPA
Health:2
Flammability:3
Instability:0
Recommended Storage : RT