missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™ Edge
Edge

For protein extraction procedures and SDS polyacrylamide gels

Supplier:  Fisher BioReagents BP166500

 View more versions of this product

Catalog No. BP166-500



Edge
Explore available promotions

Description

Description

For protein extraction procedures and SDS polyacrylamide gels

  • DNase and RNase-free detergent for protein extraction procedures and polyacrylamide gel electrophoresis
  • SDS is a commonly used detergent in protein purification and polyacrylamide gel electrophoresis (PAGE)
  • SDS denatures proteins by disrupting the non-covalent bonds of the protein
  • Functionally tested for use in electrophoresis
  • Intended for research use only; not for human or animal diagnostic purposes
Specifications

Specifications

0.1 max. (0.1% solution) at 280nm, 0.4 max. (0.1% solution) at 230nm
White
Solid
8.5 to 10
Pass Test
DNase- and RNase-Free
(as Fatty Alcohol Sulfate) >=99%
CH3(CH2)10CH2OSO3Na
15, 8768
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
3423265
288.38
Electrophoresis
Sodium Dodecyl Sulfate
151-21-3,7647-14-5,7757-82-6
206°C
500 g
Poly Bottle
(as Fatty Alcohol Sulfate) >=99%
C12H25NaO4S
MFCD00036175
Sodium Lauryl Sulfate, SDS
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984
≥99% (≥98% C12-OH)

Chemical Identifiers

C12H25NaO4S
MFCD00036175
Sodium Lauryl Sulfate, SDS
CHEBI:8984
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
3423265
sodium dodecyl sulfate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable solid. Causes eye, skin, and respiratory tract irritation. May cause allergic respiratory reaction. Avoid contact with skin and eyes. Do not breathe dust. Use explosion-proof equipment. Use only non-sparking tools. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Remove from exposure, lie down. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

Recommended Storage : RT

SDS Documents

Documents

Promotions

Promotions