missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium formate, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 446950025

 View more versions of this product

Catalog No. AC446950025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium formate
258.0°C to 261.0°C
Plastic bottle
HCO2Na
158756
Solubility in water: 550g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,insoluble in ether
C(=O)[O-].[Na+]
68.01
CHEBI:62965
ACS Reagent
141-53-7
360.0°C
CHNaO2
2.5 kg
sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
HLBBKKJFGFRGMU-UHFFFAOYSA-M
sodium;formate
2723810
68.01
0.005% max

Chemical Identifiers

141-53-7
68.01
sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
CHEBI:62965
C(=O)[O-].[Na+]
CHNaO2
HLBBKKJFGFRGMU-UHFFFAOYSA-M
2723810
sodium;formate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-488-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only