missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium fumarate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 215535000

 View more versions of this product

Catalog No. AC215535000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium fumarate
White
98%
C4H2Na2O4
MFCD00064567
02,74
disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate
VXXVUHAXJHEYFH-SEPHDYHBSA-N
(E)-but-2-enedioic acid;sodium
87110070
98%
17013-01-3
Authentic
Glass bottle
NaO2CCH=CHCO2Na
500 g
10,4165
Solubility in water: 228g/L water. Other solubilities: insoluble in ethanol, aceton
C(=CC(=O)O)C(=O)O.[Na].[Na]
160.04
160.04
Crystalline Powder

Chemical Identifiers

17013-01-3
160.04
VXXVUHAXJHEYFH-SEPHDYHBSA-N
87110070
C(=CC(=O)O)C(=O)O.[Na].[Na]
C4H2Na2O4
MFCD00064567
disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate
(E)-but-2-enedioic acid;sodium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a

GHS Signal Word: Warning

EINECSNumber : 241-087-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only