Learn More
Sodium hexafluoroaluminate, 99.9% (metals basis)
CAS: 13775-53-6 | Na3AlF6
$134.52 - $997.40
Chemical Identifiers
| CAS | 13775-53-6 |
|---|---|
| Molecular Formula | Na3AlF6 |
| MDL Number | MFCD00003507 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4447814
|
Thermo Scientific Chemicals
04447814 |
25 g |
Each for $134.52
|
|
|||||
|
AA4447822
|
Thermo Scientific Chemicals
04447822 |
100 g |
Each for $284.22
|
|
|||||
|
AA4447836
|
Thermo Scientific Chemicals
04447836 |
500 g |
Each for $997.40
|
|
|||||
Description
Sodium aluminum hexafluoride is used as a solvent (or flux) for electrolysis aluminum oxides such as bauxite, whitener for enamels and an opacifier for glass and in the industrial production of aluminum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13775-53-6 | |
| Na3AlF6 | |
| MFCD00003507 |
Specifications
| 1000°C | |
| 13775-53-6 | |
| 25 g | |
| Na3AlF6 | |
| Soluble in water(0.4g/L). | |
| Odorless | |
| Sodium hexafluoroaluminate |
| White | |
| Powder | |
| 99.9% (metals basis) | |
| MFCD00003507 | |
| 209.95 | |
| 2.9 g/cm3 at 20°C |
Safety and Handling
Do not breathe dust. Wear suitable gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.
P260-P264b-P270-P271-P304+P340-P312-P314-P501c
H332-H372
DOTInformation : DOT Class: 9; Packaging Group: III
EINECSNumber : 237-410-6
RTECSNumber : BD0075000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only