missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hexanitritocobaltate(III), ACS, Thermo Scientific Chemicals

$239.23 - $861.96

Chemical Identifiers

CAS 14649-73-1
Molecular Formula CoN6Na3O12
Molecular Weight (g/mol) 403.933
MDL Number MFCD00003512
InChI Key ZEBNDJQGEZXBCR-UHFFFAOYSA-H
Synonym cobaltic trisodium hexanitrite, trisodium cobalt 3+ hexanitrite, sodium hexanitritocobaltate 3-, cobalt 3+ sodium nitrite 1:3:6, cobaltate 3-, hexanitrito-, trisodium, cobaltate 3-, hexakis nitrito-o-, trisodium, oc-6-11, na3 co no2 6, cobaltate 3-, hexakis nitrito-kappao-, sodium 1:3 , oc-6-11
PubChem CID 159748
IUPAC Name trisodium;cobalt(3+);hexanitrite
SMILES N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[Na+].[Na+].[Na+].[Co+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1166722
View Documents
Thermo Scientific Chemicals
01166722
100 g
Each for $239.23
 
AA1166736
View Documents
Thermo Scientific Chemicals
01166736
500 g
Each for $861.96
 
Description

Description

Sodium hexanitritocobaltate(III) is used as primary and secondary intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14649-73-1
403.933
ZEBNDJQGEZXBCR-UHFFFAOYSA-H
159748
N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[Na+].[Na+].[Na+].[Co+3]
CoN6Na3O12
MFCD00003512
cobaltic trisodium hexanitrite, trisodium cobalt 3+ hexanitrite, sodium hexanitritocobaltate 3-, cobalt 3+ sodium nitrite 1:3:6, cobaltate 3-, hexanitrito-, trisodium, cobaltate 3-, hexakis nitrito-o-, trisodium, oc-6-11, na3 co no2 6, cobaltate 3-, hexakis nitrito-kappao-, sodium 1:3 , oc-6-11
trisodium;cobalt(3+);hexanitrite

Specifications

220°C (decomposition)
Yellow to Orange
100 g
MFCD00003512
14,8603
Very soluble in water
N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[Na+].[Na+].[Na+].[Co+3]
403.933
403.94
Odorless
14649-73-1
Powder
CoN6Na3O12
UN1479
cobaltic trisodium hexanitrite, trisodium cobalt 3+ hexanitrite, sodium hexanitritocobaltate 3-, cobalt 3+ sodium nitrite 1:3:6, cobaltate 3-, hexanitrito-, trisodium, cobaltate 3-, hexakis nitrito-o-, trisodium, oc-6-11, na3 co no2 6, cobaltate 3-, hexakis nitrito-kappao-, sodium 1:3 , oc-6-11
ZEBNDJQGEZXBCR-UHFFFAOYSA-H
trisodium;cobalt(3+);hexanitrite
159748
ACS
Sodium hexanitritocobaltate(III)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H334-H351-H335-H315-H319-H317
May intensify fire; oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P201-P202-P210-P220-P221-P261-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P370+P378q-P501c

H272-H315-H317-H319-H334-H335-H350

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

EINECSNumber : 237-077-7

RTECSNumber : GF9480000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only