missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium hippurate, 96%, Thermo Scientific Chemicals
$100.44 - $334.19
Chemical Identifiers
CAS | 532-94-5 |
---|---|
Molecular Formula | C9H8NNaO3 |
Molecular Weight (g/mol) | 201.157 |
MDL Number | MFCD00002693 |
InChI Key | ZBCAZEFVTIBZJS-UHFFFAOYSA-M |
Synonym | sodium hippurate, hippuric acid sodium salt, sodium 2-benzamidoacetate, n-benzoylglycine sodium salt, glycine, n-benzoyl-, monosodium salt, ccris 5812, hippuric acid, monosodium salt, glycine, n-benzoyl-, sodium salt 1:1, hippurate sodium, benzoylaminoacetic acid sodium salt |
PubChem CID | 516953 |
IUPAC Name | sodium;2-benzamidoacetate |
SMILES | C1=CC=C(C=C1)C(=O)NCC(=O)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1172318
|
Thermo Scientific Chemicals
A1172318 |
50 g |
Each for $100.44
|
|
|||||
AAA1172330
|
Thermo Scientific Chemicals
A1172330 |
250 g |
Each for $334.19
|
|
|||||
Description
It may be a useful urinary biomarker for certain cancers such as gastric cancer and oxidative stress.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt may be a useful urinary biomarker for certain cancers such as gastric cancer and oxidative stress.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
532-94-5 | |
201.157 | |
ZBCAZEFVTIBZJS-UHFFFAOYSA-M | |
516953 | |
C1=CC=C(C=C1)C(=O)NCC(=O)[O-].[Na+] |
C9H8NNaO3 | |
MFCD00002693 | |
sodium hippurate, hippuric acid sodium salt, sodium 2-benzamidoacetate, n-benzoylglycine sodium salt, glycine, n-benzoyl-, monosodium salt, ccris 5812, hippuric acid, monosodium salt, glycine, n-benzoyl-, sodium salt 1:1, hippurate sodium, benzoylaminoacetic acid sodium salt | |
sodium;2-benzamidoacetate |
Specifications
532-94-5 | |
C9H8NNaO3 | |
50 g | |
Soluble in water. | |
C1=CC=C(C=C1)C(=O)NCC(=O)[O-].[Na+] | |
201.157 | |
201.16 | |
Powder or Lumps |
96% | |
MFCD00002693 | |
sodium hippurate, hippuric acid sodium salt, sodium 2-benzamidoacetate, n-benzoylglycine sodium salt, glycine, n-benzoyl-, monosodium salt, ccris 5812, hippuric acid, monosodium salt, glycine, n-benzoyl-, sodium salt 1:1, hippurate sodium, benzoylaminoacetic acid sodium salt | |
ZBCAZEFVTIBZJS-UHFFFAOYSA-M | |
sodium;2-benzamidoacetate | |
516953 | |
96% | |
Sodium Hippurate |
Safety and Handling
EINECSNumber : 208-548-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only