missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydrogen DL-malate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1942818

 View more versions of this product

Catalog No. AAA1942818

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium hydrogen DL-malate
210°C (decomposition)
MFCD00798569
sodium hydrogen malate, monosodium malate, sodium malate, l-malic acid, --sodium hydrogen malate, l--malic acid disodium salt, sodium 3-carboxy-3-hydroxypropanoate, 2-hydroxysuccinic acid 1-sodium salt, sodium 3,4-dihydroxy-4-oxo-butanoate, 2-hydroxybutanedioic acid 4-sodium salt
[Na+].OC(CC([O-])=O)C(O)=O
156.07
156.07
57467-17-1
C4H5NaO5
50 g
DOJOZCIMYABYPO-UHFFFAOYNA-M
sodium;3,4-dihydroxy-4-oxobutanoate
23678821
98%

Chemical Identifiers

57467-17-1
156.07
DOJOZCIMYABYPO-UHFFFAOYNA-M
23678821
[Na+].OC(CC([O-])=O)C(O)=O
C4H5NaO5
MFCD00798569
sodium hydrogen malate, monosodium malate, sodium malate, l-malic acid, --sodium hydrogen malate, l--malic acid disodium salt, sodium 3-carboxy-3-hydroxypropanoate, 2-hydroxysuccinic acid 1-sodium salt, sodium 3,4-dihydroxy-4-oxo-butanoate, 2-hydroxybutanedioic acid 4-sodium salt
sodium;3,4-dihydroxy-4-oxobutanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only