missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium hydrogen tartrate, 98+%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 389422500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium hydrogen tartrate | |
White | |
3.3 to 3.6 (5% soln.) | |
250 g | |
C4H5NaO6 | |
15, 8725 | |
Solubility in water: 89g/L (20°C) | |
[Na+].OC(C(O)C([O-])=O)C(O)=O | |
172.07 | |
172.07 | |
Analytical |
253°C | |
526-94-3 | |
Powder | |
98+% | |
MFCD00065393,MFCD00065393 | |
sodium bitartrate, monosodium tartrate, sodium hydrogen tartrate, natriumtartrat german, sodium 3-carboxy-2,3-dihydroxypropanoate, monobasic sodium tartrate, monosodium l-+-tartrate, tartaric acid, monosodium salt, natrium hydrogen-2r,3r-tartrat, weinstein | |
NKAAEMMYHLFEFN-UHFFFAOYNA-M | |
sodium;2,3,4-trihydroxy-4-oxobutanoate | |
23690454 | |
≥98% | |
Plastic bottle |
Chemical Identifiers
526-94-3 | |
172.07 | |
NKAAEMMYHLFEFN-UHFFFAOYNA-M | |
23690454 | |
[Na+].OC(C(O)C([O-])=O)C(O)=O |
C4H5NaO6 | |
MFCD00065393,MFCD00065393 | |
sodium bitartrate, monosodium tartrate, sodium hydrogen tartrate, natriumtartrat german, sodium 3-carboxy-2,3-dihydroxypropanoate, monobasic sodium tartrate, monosodium l-+-tartrate, tartaric acid, monosodium salt, natrium hydrogen-2r,3r-tartrat, weinstein | |
sodium;2,3,4-trihydroxy-4-oxobutanoate |
Safety and Handling
EINECSNumber : 208-400-9
RUO – Research Use Only