missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hydroxide, 50% (w/v) Solution, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S22420LTCU

 View more versions of this product

Catalog No. 18613614


Explore available promotions

Description

Description

Specifications

Specifications

1310-73-2,7732-18-5
50% ±1%
Cubitainer
20 L
Chemical Solution

Chemical Identifiers

1310-73-2
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.